Order Entry
Export
Orders LinkContactUsLinkComponent
7-Hydroxycoumarin 99%

7-Hydroxycoumarin 99%

Plastic bottle, 100 g
Catalog # ACRO121111000
7-Hydroxycoumarin 99%
Catalog # ACRO121111000
CAS Number:  93-35-6
7-Hydroxycoumarin 99%
7-Hydroxycoumarin 99%

Plastic bottle, 100 g

CAS Number:  93-35-6
Price
152,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • CAS Number:93-35-6
  • Formula:C₉H₆O₃
  • MDL Number:MFCD00006878
  • Molecular Weight:162.14 g/mol
  • Merck Index:12,09981
  • Boiling Pt:382 °C (1013 hPa)
  • Melting Pt:228…234 °C
  • Storage Temperature:Ambient
  • EINECS:202-240-3

Documents

Specifications

Specification Test Results

  • Appearance:
    White to yellow to beige to light brown Powder or crystals and/or agglomerates
  • Infrared spectrum:
    Conforms
  • Melting point:
    228 to 234 °C
  • HPLC:
    ≥98.5 %
  • Titration with NaOH:
    98.5 to 101.5 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications