7-Hydroxycoumarin 99%
Plastic bottle, 25 g
: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
- :93-35-6
- :C₉H₆O₃
- :MFCD00006878
- :162.14 g/mol
- :12,09981
- :382 °C (1013 hPa)
- :228…234 °C
- :Ambient
- :202-240-3
- Appearance:White to yellow to beige to light brown Powder or crystals and/or agglomerates
- Infrared spectrum:Conforms
- Melting point:228 to 234 °C
- HPLC:≥98.5 %
- Titration with NaOH:98.5 to 101.5 %
- Pack type:Plastic bottle
- Pk:25 g



