1-Hexanol 98%, pure
Glass bottle, 2,5 L
: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
- :111-27-3
- :H₃C(CH₂)₄CH₂OH
- :MFCD00002982
- :102.18 g/mol
- :13,04715
- :156…157 °C (760 mmHg)
- :–52 °C
- :0.8136 g/cm³
- :59 °C
- :Ambient
- :203-852-3
- :2282
- :3,III
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4170 to 1.4190 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥97.5 %
- Color scale:≤10 APHA
- Acid value:≤0.02 mg KOH/g
- Ester value:≤0.3 mg KOH/g
- Iodine value:≤0.1 g I2/100 g
- Pack type:Glass bottle
- Pk:2,5 L



