1-Hexanol 98%, pure
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
- CAS Number:111-27-3
- Formula:H₃C(CH₂)₄CH₂OH
- MDL Number:MFCD00002982
- Molecular Weight:102.18 g/mol
- Merck Index:13,04715
- Boiling Pt:156…157 °C (760 mmHg)
- Melting Pt:–52 °C
- Density:0.8136 g/cm³
- Flash Pt:59 °C
- Storage Temperature:Ambient
- EINECS:203-852-3
- UN:2282
- ADR:3,III
Documents
ACRO12079
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4170 to 1.4190 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥97.5 %
- Color scale:≤10 APHA
- Acid value:≤0.02 mg KOH/g
- Ester value:≤0.3 mg KOH/g
- Iodine value:≤0.1 g I2/100 g
Specifications
- Pack type:Glass bottle
- Pk:1 L



