Order Entry
Export
Orders LinkContactUsLinkComponent
1,6-Hexanediol 97%

1,6-Hexanediol 97%

Glass bottle for solids, 2,5 kg
Catalog # ACRO120650025
1,6-Hexanediol 97%
Catalog # ACRO120650025
CAS Number:  629-11-8
1,6-Hexanediol 97%
1,6-Hexanediol 97%

Glass bottle for solids, 2,5 kg

CAS Number:  629-11-8
Price
147,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Hexamethylene glycol, 1,6-Dihydroxy hexane
  • CAS Number:629-11-8
  • Formula:C₆H₁₄O₂
  • MDL Number:MFCD00002985
  • Molecular Weight:118.18 g/mol
  • Merck Index:13,04709
  • Boiling Pt:253…260 °C (760 mmHg)
  • Melting Pt:40…43 °C
  • Density:0.96 g/cm³
  • Flash Pt:147 °C
  • Storage Temperature:Ambient
  • EINECS:211-074-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Colorless to white to light yellow Solidified melt or liquid
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 44 °C
  • Water:
    ≤0.3 % (K.F.)
  • GC:
    ≥96.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications