Order Entry
Export
Orders LinkContactUsLinkComponent
Guanine 99+%

Guanine 99+%

Glass bottle for solids, 10 g
Catalog # ACRO120250100
Irritant
Guanine 99+%
Catalog # ACRO120250100
CAS Number:  73-40-5
Guanine 99+%
Guanine 99+%

Glass bottle for solids, 10 g

CAS Number:  73-40-5
Price
33,90 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  2-Amino-1H-purin-6(7H)-one
  • CAS Number:73-40-5
  • Formula:C₅H₅N₅O
  • MDL Number:MFCD00071533
  • Molecular Weight:151.13 g/mol
  • Merck Index:13,04580
  • Melting Pt:360 °C (dec.)
  • Density:1,67 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-799-8

Documents

Specifications

Specification Test Results

  • Appearance:
    White to cream Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥99.0 %
  • Residue after ignition:
    ≤0.1 %
  • Ratio of Absorbances:
    1.35 to 1.41 (A250/A260)
  • Titration with HClO4:
    ≥99.0 % (On dry substance)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications