Order Entry
Export
Orders LinkContactUsLinkComponent
3-Fluoroaniline 98%

3-Fluoroaniline 98%

Glass bottle, 25 mL
Catalog # ACRO119280250
Irritant
3-Fluoroaniline 98%
Catalog # ACRO119280250
CAS Number:  372-19-0
3-Fluoroaniline 98%
3-Fluoroaniline 98%

Glass bottle, 25 mL

CAS Number:  372-19-0
Price
57,10 €
Log in for availability
1 * 25 mL
Product Details & Documents
  • CAS Number:372-19-0
  • Formula:C₆H₆FN
  • MDL Number:MFCD00007758
  • Molecular Weight:111.12 g/mol
  • Boiling Pt:186 °C (756 mmHg)
  • Melting Pt:–2 °C
  • Density:1.15 g/cm³
  • Flash Pt:77 °C
  • Storage Temperature:Ambient
  • EINECS:206-747-0
  • UN:2941
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5430 to 1.5450 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL

Specifications