Ethyl-(S)-(-)-lactate 97%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Ethyl-(S)-(-)-lactate 97%
Catalog # ACRO118660010
Supplier: Thermo Fisher Scientific
CAS Number: 687-47-8
Jump to:
Product Details & Documents
Synonyms: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate
- CAS Number:687-47-8
- Formula:C₅H₁₀O₃
- MDL Number:MFCD00004518
- Molecular Weight:118.13 g/mol
- Merck Index:12,03850
- Boiling Pt:154 °C (760 mmHg)
- Melting Pt:–26 °C
- Density:1.034 g/cm³
- Flash Pt:46 °C
- Storage Temperature:Ambient
- EINECS:211-694-1
- UN:1192
- ADR:3,III
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4120 to 1.4140 (20°C, 589 nm)
- Water:≤0.3 % (K.F.)
- GC:≥96.0 %
- Color scale:≤50 APHA
- Specific gravity:(20°C) 1.03 to 1.038
- Non-volatile matter:≤0.01 %
- Specific optical rotation:-8° to -12° (20°C, 589 nm) (neat)
- Acidity:≤0.1 % (as lactic acid)
Specifications
- Pack type:Glass bottle
- Pk:1 L



