1,1-Diphenylethylene 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
1,1-Diphenylethylene 98%
Catalog # ACRO117260250
Supplier: Thermo Fisher Scientific
CAS Number: 530-48-3
Jump to:
Product Details & Documents
Synonyms: asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
- CAS Number:530-48-3
- Formula:(C₆H₅)₂CCH₂
- MDL Number:MFCD00008583
- Molecular Weight:180.25 g/mol
- Merck Index:13,03355
- Boiling Pt:270…271 °C (760 mmHg)
- Melting Pt:6 °C
- Density:1.02 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Refrigerator
- EINECS:208-482-6
Specifications
Specification Test Results
- Appearance:Clear colorless to golden Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6070 to 1.6090 (20°C, 589 nm)
- GC:≥97.5 %
- Iodide (I):≤30 ppm
- Phosphate (PO4):≤10 ppm
- SO4 and SO3 (as SO4):≤0.003 %
- Iron (Fe):≤1 ppm
- Selenium (Se):≤0.01 ppm
- Heavy metals (ICP-OES):≤5 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



