1,1-Diphenylethylene 98%
Glass bottle for solids, 10 g
: asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
- :530-48-3
- :(C₆H₅)₂CCH₂
- :MFCD00008583
- :180.25 g/mol
- :13,03355
- :270…271 °C (760 mmHg)
- :6 °C
- :1.02 g/cm³
- :>112 °C
- :Refrigerator
- :208-482-6
- Appearance:Clear colorless to golden Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6070 to 1.6090 (20°C, 589 nm)
- GC:≥97.5 %
- Iodide (I):≤30 ppm
- Phosphate (PO4):≤10 ppm
- SO4 and SO3 (as SO4):≤0.003 %
- Iron (Fe):≤1 ppm
- Selenium (Se):≤0.01 ppm
- Heavy metals (ICP-OES):≤5 ppm
- Pack type:Glass bottle for solids
- Pk:10 g



