1,4-Dioxane 99+% stabilised, extra pure
Metal drum, 25 L
Supplier: Thermo Fisher Scientific
1,4-Dioxane 99+% stabilised, extra pure
Catalog # ACRO117110250
Supplier: Thermo Fisher Scientific
CAS Number: 123-91-1
Jump to:
Product Details & Documents
Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
- CAS Number:123-91-1
- Formula:C₄H₈O₂
- MDL Number:MFCD00006571
- Molecular Weight:88.11 g/mol
- Merck Index:13,03330
- Boiling Pt:101.1 °C (1013 hPa)
- Melting Pt:12 °C
- Density:1.034 g/cm³
- Storage Temperature:Ambient
- EINECS:204-661-8
- UN:1165
- ADR:3,II
Documents
ACRO11711
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.0 %
- Color scale:≤20 APHA
- Stabilizer:5 to 30 ppm BHT
- Residue after evaporation:≤0.01 %
- Peroxides:≤0.005 % (as H2O2)
Specifications
- Pack type:Metal drum
- Pk:25 L



