1,4-Dioxane 99+% stabilised, extra pure
Glass bottle, 1 L
: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
- :123-91-1
- :C₄H₈O₂
- :MFCD00006571
- :88.11 g/mol
- :13,03330
- :101.1 °C (1013 hPa)
- :12 °C
- :1.034 g/cm³
- :Ambient
- :204-661-8
- :1165
- :3,II
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.0 %
- Color scale:≤20 APHA
- Stabilizer:5 to 30 ppm BHT
- Residue after evaporation:≤0.01 %
- Peroxides:≤0.005 % (as H2O2)
- Pack type:Glass bottle
- Pk:1 L



