3-(Dimethylamino)phenol 97%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
3-(Dimethylamino)phenol 97%
Catalog # ACRO115861000
Supplier: Thermo Fisher Scientific
CAS Number: 99-07-0
Jump to:
Product Details & Documents
Synonyms: N,N-Dimethyl-3-hydroxyaniline, 3-Dimethylaminophenol
- CAS Number:99-07-0
- Formula:C₈H₁₁NO
- MDL Number:MFCD00002264
- Molecular Weight:137.18 g/mol
- Boiling Pt:265 °C (1013 hPa)
- Melting Pt:80…85 °C
- Flash Pt:148 °C
- Storage Temperature:Ambient
- EINECS:202-727-0
Documents
ACRO11586
SDS
Specifications
Specification Test Results
- Appearance:Grey to brown to purple Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Water:≤0.05 % (Coulometric)
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Substance darkened by H2SO4:Passes test
- Sulfur compounds (as S):≤0.003 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



