Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,5-Dimethoxybenzaldehyde 97%

Glass bottle for solids, 25 g
  ACRO115390250
 
2,5-Dimethoxybenzaldehyde 97%
  ACRO115390250
 :  93-02-7
2,5-Dimethoxybenzaldehyde 97%
2,5-Dimethoxybenzaldehyde 97%

Glass bottle for solids, 25 g

 :  93-02-7
 
31,20 €
   
1 * 25 g
 
  •  :93-02-7
  •  :C₉H₁₀O₃
  •  :MFCD00003314
  •  :166.18 g/mol
  •  :146 °C (10 mmHg)
  •  :45…50 °C
  •  :113 °C
  •  :Ambient
  •  :202-211-5

 

 

 

  • Appearance:
    Yellow to beige Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    46 to 51 °C
  • Water:
    ≤0.5 %
  • GC:
    ≥96.0 %

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g