1,5-Dihydroxynaphthalene 97%
Plastic bottle, 500 g
: 1,5-Naphthalenediol, Naphthalene-1,5-diol
- :83-56-7
- :C₁₀H₈O₂
- :MFCD00003980
- :160.17 g/mol
- :375 °C (1013 hPa)
- :259…261 °C (dec.)
- :1,41 g/cm³ (20 °C)
- :252 °C
- :Ambient
- :201-487-4
- :3077
- :9,III
- Appearance:Grey-brown to brown Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Pack type:Plastic bottle
- Pk:500 g



