1,5-Dihydroxynaphthalene 97%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
1,5-Dihydroxynaphthalene 97%
Catalog # ACRO115001000
Supplier: Thermo Fisher Scientific
CAS Number: 83-56-7
Jump to:
Product Details & Documents
Synonyms: 1,5-Naphthalenediol, Naphthalene-1,5-diol
- CAS Number:83-56-7
- Formula:C₁₀H₈O₂
- MDL Number:MFCD00003980
- Molecular Weight:160.17 g/mol
- Boiling Pt:375 °C (1013 hPa)
- Melting Pt:259…261 °C (dec.)
- Density:1,41 g/cm³ (20 °C)
- Flash Pt:252 °C
- Storage Temperature:Ambient
- EINECS:201-487-4
- UN:3077
- ADR:9,III
Documents
ACRO11500
SDS
Specifications
Specification Test Results
- Appearance:Grey-brown to brown Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specifications
- Pack type:Plastic bottle
- Pk:100 g



