2,5-Dihydroxybenzaldehyde 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
2,5-Dihydroxybenzaldehyde 99%
Catalog # ACRO114840050
Supplier: Thermo Fisher Scientific
CAS Number: 1194-98-5
Jump to:
Product Details & Documents
- CAS Number:1194-98-5
- Formula:C₇H₆O₃
- MDL Number:MFCD00003333
- Molecular Weight:138.12 g/mol
- Boiling Pt:60 - 65 °C (12 -rr)
- Melting Pt:98…102 °C
- Density:0,859 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:214-789-6
Specifications
Specification Test Results
- Appearance:Yellow to khaki Crystalline powder
- Infrared spectrum:Conforms
- Melting point:98 to 102 °C
- GC:≥98.5 %
- Copper (Cu):≤50 ppm
- Iron (Fe):≤100 ppm
- Lead (Pb):≤50 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



