Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl phthalate 99%

Diethyl phthalate 99%

Glass bottle, 1 L
Catalog # ACRO114520010
Diethyl phthalate 99%
Catalog # ACRO114520010
CAS Number:  84-66-2
Diethyl phthalate 99%
Diethyl phthalate 99%

Glass bottle, 1 L

CAS Number:  84-66-2
Price
51,70 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Phthalic acid diethyl ester
  • CAS Number:84-66-2
  • Formula:C₁₂H₁₄O₄
  • MDL Number:MFCD00009111
  • Molecular Weight:222.24 g/mol
  • Merck Index:12,07527
  • Boiling Pt:298…299 °C (760 mmHg)
  • Melting Pt:-3 °C
  • Density:1.118 g/cm³
  • Flash Pt:156 °C
  • Storage Temperature:Ambient
  • EINECS:201-550-6

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5010 to 1.5030 (20°C, 589 nm)
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤25 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications