Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,6-Dibromohexane 98%

Glass bottle, 100 mL
Catalog # ACRO112811000
Irritant
Environment
1,6-Dibromohexane 98%
Catalog # ACRO112811000
CAS Number:  629-03-8
1,6-Dibromohexane 98%
1,6-Dibromohexane 98%

Glass bottle, 100 mL

CAS Number:  629-03-8
Price
38,50 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Hexamethylen dibromide
  • CAS Number:629-03-8
  • Formula:BrH₂C(CH₂)₄CH₂Br
  • MDL Number:MFCD00000272
  • Molecular Weight:243.97 g/mol
  • Boiling Pt:243 °C (760 mmHg)
  • Melting Pt:–2 °C
  • Density:1.58 g/cm³
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • EINECS:211-067-2
  • UN:3082
  • ADR:9,III

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5056 to 1.5076 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    +29° to +33° (20°C, 589 nm) (c=1, toluene)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications