1,2-Dibromoethane 99%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
1,2-Dibromoethane 99%
Catalog # ACRO112790010
Supplier: Thermo Fisher Scientific
CAS Number: 106-93-4
Jump to:
Product Details & Documents
- CAS Number:106-93-4
- Formula:BrH₂CCH₂Br
- MDL Number:MFCD00000233
- Molecular Weight:187.86 g/mol
- Merck Index:13,03830
- Boiling Pt:131…132°C (760mmHg)
- Boiling Pt:131…132 °C (760 mmHg)
- Melting Pt:9…10°C
- Melting Pt:9…10 °C
- Density:2.173
- Flash Pt:>104°C
- Flash Pt:>104 °C
- Storage Temperature:Ambient
- EINECS:203-444-5
- UN:1605
- ADR:6.1,I
- ADR:6.1,
Documents
ACRO11279
SDS
Specifications
Specification Test Results
- Appearance:clear colorless to light yellow liquid
- Infrared spectrum:Conforms
- GC:≥98.5%
- Refractive index:1.5375 to 1.5395 (20°C, 589nm)
Specifications
- Pack type:Glass bottle
- Pk:1 L



