1,5-Diaminonaphthalene 97%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
1,5-Diaminonaphthalene 97%
Catalog # ACRO112291000
Supplier: Thermo Fisher Scientific
CAS Number: 2243-62-1
Jump to:
Product Details & Documents
Synonyms: 1,5-naphthylenediamine, 1,5-Naphthalenediamine
- CAS Number:2243-62-1
- Formula:C₁₀H₁₀N₂
- MDL Number:MFCD00004029
- Molecular Weight:158.2 g/mol
- Boiling Pt:381 °C (1013 hPa)
- Melting Pt:187…190 °C
- Density:1,4 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:218-817-8
- UN:3077
- ADR:9,III
Specifications
Specification Test Results
- Appearance:Grey to dark brown Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Titration with HClO4:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



