1,12-Dodecanedioic acid 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
1,12-Dodecanedioic acid 99%
Catalog # ACRO111880050
Supplier: Thermo Fisher Scientific
CAS Number: 693-23-2
Jump to:
Product Details & Documents
Synonyms: Dodecanedioic acid, 1,10-Decanedicarboxylic acid
- CAS Number:693-23-2
- Formula:C₁₂H₂₂O₄
- MDL Number:MFCD00002735
- Molecular Weight:230.3 g/mol
- Boiling Pt:245 °C (10 mmHg)
- Melting Pt:125…130 °C
- Density:1,175 g/cm³ (21 °C)
- Flash Pt:220 °C
- Storage Temperature:Ambient
- EINECS:211-746-3
Specifications
Specification Test Results
- Appearance:White Powder or pellets
- Infrared spectrum:Conforms
- Melting point:125 to 130 °C
- Titration with NaOH:≥98.5 %
- Viscosity:30.5 to 36.5
- pH:3.0 to 3.8 (1 % solution)
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



