n-Decane 99+%, pure
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:124-18-5
- Formula:H₃C(CH₂)₈CH₃
- MDL Number:MFCD00008954
- Molecular Weight:142.28 g/mol
- Boiling Pt:174 °C (760 mmHg)
- Melting Pt:–30 °C
- Density:0.735 g/cm³
- Flash Pt:46 °C
- Storage Temperature:Ambient
- EINECS:204-686-4
- UN:2247
- ADR:3,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4100 to 1.4130 (20°C, 589 nm)
- GC:≥99.0 %
- Composition of fatty acids:5.0 to 14.0 % (GC) Palmitic acid
- Acid value:≤0.5 mg KOH/g
- Peroxide value:≤5 meq O2/kg
- Unsaponifiable matter:≤1 %
Specifications
- Pack type:Glass bottle
- Pk:500 mL



