m-Cresol 99%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene
- CAS Number:108-39-4
- Formula:CH₃C₆H₄OH
- MDL Number:MFCD00002302
- Molecular Weight:108.14 g/mol
- Merck Index:13,02604
- Boiling Pt:203 °C (760 mmHg)
- Melting Pt:8…10 °C
- Density:1.03 g/cm³
- Flash Pt:86 °C
- Storage Temperature:Ambient
- EINECS:203-577-9
- UN:2076
- ADR:6.1,II
Specifications
Specification Test Results
- Appearance:Clear colorless to light red Viscous liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5390 to 1.5420 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



