Coronene 95%
Glass bottle for solids, 500 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Hexabenzobenzene
- CAS Number:191-07-1
- Formula:C₂₄H₁₂
- MDL Number:MFCD00004134
- Molecular Weight:300.36 g/mol
- Boiling Pt:525 °C (1013 hPa)
- Melting Pt:438…440 °C
- Density:1,371 g/cm³ (25 °C)
- EINECS:205-881-7
- UN:2811
Documents
ACRO11051
SDS
Specifications
Specification Test Results
- Appearance:Yellow to green Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Titration RCOCl:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



