(±)-β-Citronellol 95%
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
(±)-β-Citronellol 95%
Catalog # ACRO110461000
Supplier: Thermo Fisher Scientific
CAS Number: 106-22-9
Jump to:
Product Details & Documents
Synonyms: DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol
- CAS Number:106-22-9
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00002935
- Molecular Weight:156.27 g/mol
- Merck Index:13,02354
- Boiling Pt:222 °C (1013 hPa)
- Density:0.854 g/cm³
- Flash Pt:79 °C
- EINECS:203-375-0
- ADR:9,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4540 to 1.4580 (20°C, 589 nm)
- GC:≥94 %
- Specific gravity:(25°C) 0.850 to 0.860
Specifications
- Pack type:Glass bottle
- Pk:100 mL



