Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-β-Citronellol 95%

(±)-β-Citronellol 95%

Glass bottle, 5 mL
Catalog # ACRO110460050
Irritant
(±)-β-Citronellol 95%
Catalog # ACRO110460050
CAS Number:  106-22-9
(±)-β-Citronellol 95%
(±)-β-Citronellol 95%

Glass bottle, 5 mL

CAS Number:  106-22-9
Price
28,40 €
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol
  • CAS Number:106-22-9
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00002935
  • Molecular Weight:156.27 g/mol
  • Merck Index:13,02354
  • Boiling Pt:222 °C (1013 hPa)
  • Density:0.854 g/cm³
  • Flash Pt:79 °C
  • EINECS:203-375-0
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4540 to 1.4580 (20°C, 589 nm)
  • GC:
    ≥94 %
  • Specific gravity:
    (25°C) 0.850 to 0.860

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications