Order Entry
Export
Orders LinkContactUsLinkComponent
DNA modification enzymes

DNA modification enzymes

500 EU
Catalog # E70092Y
Supplier:   Cytiva
DNA modification enzymes
Catalog # E70092Y
Supplier:  Cytiva
CAS Number:  
DNA modification enzymes
DNA modification enzymes

500 EU

CAS Number:  
Price
138,00 €
Log in for availability
1 * 500 EU
Product Details & Documents

A high specific activity, heat-labile alkaline phosphatase. Completely and irreversibly inactivated in Tris buffers at pH 8,0 to 8,5 by heating for 15 minutes at 65 °C. No further treatment is necessary.

  • Bacterial alkaline phosphatase recombinantly expressed in E coli
  • Active in PCR and restriction endonuclease buffers
  • Inactivated by heating at 75°C for 5 minutes
  • Easily soluble in cold water and hot water
  • Sourced from arctic shrimp (Pandalus borealis)

Use together with illustra Exonuclease I for cleaning up PCR reactions prior to downstream application such as DNA sequencing. Dephosphorylation of a restriction enzyme digested plasmid (5-20 pmol of 5'-ends, 0.1-0.5 units/pmol 5'-ends). Reduces re-ligation to < 0.5% compared to the untreated control.

One unit is the amount of the enzyme required to dephosphorylate 1 µg of linearized pUC57 DNA termini in 10 minutes at 37°C. The enzyme is purified to apparent homogeneity and is free of all contaminating endonucleases, exonucleases and ribonucleases

Unit Definition: One unit is the amount of the enzyme required to dephosphorylate 1 Â µg of linearized pUC57 DNA termini in 10 minutes at 37 °C.

Delivery information: Includes 10X reaction buffer: 200 mM Tris-HCl, pH 8,0, 100 mM MgCl₂; dilution buffer: 50 mM Tris-HCl, pH 8,0.

Specifications

Specifications

  • Pk:
    500 EU
  • Cat. No.:
    E70092Y
  • Enzyme activity:
    1 unit/µl
  • Purity:
    The enzyme is purified to apparent homogeneity and is free of all contaminating endonucleases, exonucleases and ribonucleases
  • Enzyme name:
    DNA modification enzymes

Specifications