2-Bromotoluene 99%
Glass bottle, 25 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:95-46-5
- Formula:C₇H₇Br
- MDL Number:MFCD00000068
- Molecular Weight:171.04 g/mol
- Merck Index:13,01424
- Boiling Pt:181.5 °C (1013 hPa)
- Melting Pt:–27 °C
- Density:1.42 g/cm³
- Flash Pt:78 °C
- Storage Temperature:Refrigerator
- EINECS:202-421-7
- ADR:9,III
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5550 to 1.5570 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 mL



