Order Entry
Export
Orders LinkContactUsLinkComponent
2-Bromopyridine 99%

2-Bromopyridine 99%

Glass bottle, 25 mL
Catalog # ACRO107400250
Toxic
Flammable
2-Bromopyridine 99%
Catalog # ACRO107400250
CAS Number:  109-04-6
2-Bromopyridine 99%
2-Bromopyridine 99%

Glass bottle, 25 mL

CAS Number:  109-04-6
Price
41,40 €
Log in for availability
1 * 25 mL
Product Details & Documents
  • CAS Number:109-04-6
  • Formula:C₅H₄BrN
  • MDL Number:MFCD00006219
  • Molecular Weight:158 g/mol
  • Boiling Pt:192…194 °C (761 mmHg)
  • Melting Pt:-40,1 °C
  • Density:1.65 g/cm³
  • Flash Pt:54 °C
  • Storage Temperature:Ambient
  • EINECS:203-641-6
  • UN:2929
  • ADR:6.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5700 to 1.5730 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤20 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL

Specifications