Order Entry
Export
Orders LinkContactUsLinkComponent
4-Phenylbenzaldehyde 99%

4-Phenylbenzaldehyde 99%

Glass bottle for solids, 25 g
Catalog # ACRO106260250
Irritant
4-Phenylbenzaldehyde 99%
Catalog # ACRO106260250
CAS Number:  3218-36-8
4-Phenylbenzaldehyde 99%
4-Phenylbenzaldehyde 99%

Glass bottle for solids, 25 g

CAS Number:  3218-36-8
Price
84,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
  • CAS Number:3218-36-8
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00006947
  • Molecular Weight:182.22 g/mol
  • Boiling Pt:184 °C (11 mmHg)
  • Melting Pt:57…60 °C
  • Storage Temperature:Ambient
  • EINECS:221-742-3

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    57 to 60 °C
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications