Order Entry
Export
Orders LinkContactUsLinkComponent
Benzil 99+%

Benzil 99+%

Plastic bottle, 500 g
Catalog # ACRO105415000
Irritant
Benzil 99+%
Catalog # ACRO105415000
CAS Number:  134-81-6
Benzil 99+%
Benzil 99+%

Plastic bottle, 500 g

CAS Number:  134-81-6
Price
74,40 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1,2-Diphenyl-1,2-ethanedione
  • CAS Number:134-81-6
  • Formula:(C₆H₅)C(O)C(O)(C₆H₅)
  • MDL Number:MFCD00003080
  • Molecular Weight:210.23 g/mol
  • Merck Index:13,01078
  • Boiling Pt:346…348 °C (760 mmHg)
  • Melting Pt:94…97 °C
  • Density:1,23 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-157-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    93 to 97 °C
  • GC:
    ≥99.0 %
  • Calcium (Ca):
    ≤100 ppm
  • Iron (Fe):
    ≤500 ppm
  • Potassium (K):
    ≤200 ppm
  • Sodium (Na):
    ≤500 ppm
  • Carbon compounds (as C):
    ≤0.01 %
  • Insoluble matter:
    ≤0.02 % (in dilute HCl)
  • Nitrogen compounds (as N):
    ≤0.002 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications