Anthranilic acid 98+%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Anthranilic acid 98+%
Catalog # ACRO104915000
Supplier: Thermo Fisher Scientific
CAS Number: 118-92-3
Jump to:
Product Details & Documents
Synonyms: 2-Aminobenzoic acid
- CAS Number:118-92-3
- Formula:C₇H₇NO₂
- MDL Number:MFCD00007712
- Molecular Weight:137.14 g/mol
- Merck Index:13,00421
- Boiling Pt:312 °C (1013 hPa)
- Melting Pt:144…148 °C
- Density:1.41 g/cm³
- Flash Pt:150 °C
- :Density: 1.362 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-287-5
Specifications
Specification Test Results
- Appearance:Light yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:144 to 148 °C
- HPLC:≥98.0 %
- Free amine:≤0.01 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g



