Anthranilic acid 98+%
Plastic bottle, 100 g
: 2-Aminobenzoic acid
- :118-92-3
- :C₇H₇NO₂
- :MFCD00007712
- :137.14 g/mol
- :13,00421
- :312 °C (1013 hPa)
- :144…148 °C
- :1.41 g/cm³
- :150 °C
- :Density: 1.362 g/cm³ (20 °C)
- :Ambient
- :204-287-5
- Appearance:Light yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:144 to 148 °C
- HPLC:≥98.0 %
- Free amine:≤0.01 %
- Pack type:Plastic bottle
- Pk:100 g



