Orders LinkContactUsLinkComponent
Accessories for chromatography systems
Union, M6 female for tubing ¹/₁₆", PEEK, 2
Catalog # 18-1123-94
Supplier:   Cytiva
Accessories for chromatography systems
Catalog # 18-1123-94
Supplier:  Cytiva
CAS Number:  
Accessories for chromatography systems
Accessories for chromatography systems
Union, M6 female for tubing ¹/₁₆", PEEK, 2
CAS Number:  
Price
156,00 €
Log in for availability
1 * 2 items
Specifications
Specifications
  • Pk:
    2
  • Description:
    Union, M6 female for tubing ¹/₁₆", PEEK
Specifications