Order Entry
Export
Orders LinkContactUsLinkComponent
Accessories for chromatography systems

Accessories for chromatography systems

Inlet tubing kit, FEP, Int.Ø 2,9 mm, Ext.Ø 4,76 mm, 1
Catalog # 18-1114-40
Supplier:   Cytiva
Accessories for chromatography systems
Catalog # 18-1114-40
Supplier:  Cytiva
CAS Number:  
Accessories for chromatography systems
Accessories for chromatography systems

Inlet tubing kit, FEP, Int.Ø 2,9 mm, Ext.Ø 4,76 mm, 1

CAS Number:  
Price
181,00 €
Log in for availability
1 * 1 items
Specifications

Specifications

  • Pk:
    1
  • Description:
    Inlet tubing kit, FEP, Int.Ø 2,9 mm, Ext.Ø 4,76 mm

Specifications