Paracetamol 98%
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
- CAS Number:103-90-2
- Formula:C₈H₉NO₂
- MDL Number:MFCD00002328
- Molecular Weight:151.16 g/mol
- Merck Index:13,00048
- Boiling Pt:388 °C (1013 hPa)
- Melting Pt:168…172 °C
- Density:1.293 g/cm³
- Storage Temperature:Ambient
- EINECS:203-157-5
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:168 to 172 °C
- HPLC:≥97.5 %
- Gel strength:500 to 1000 g/cm² (1.5 % at 20 °C)
- Ash:≤6 %
- Microbial contamination:≤1000 /g (Total plate count)
- pH:5.7 to 7.5 (1.5 % in water)
Specifications
- Pack type:Plastic bottle
- Pk:250 g



