ContactUsLinkComponent
Thermometer, AMA freezer, red alcohol filled, measuring range:-35:20/0.5C, capillary yellow backed, dimension: 280x28mm, enclosed s cale, complete in plastic casing, rounded top finish, government verified with verification stamp. 1 * 1 items
Catalog # 620-1053
Supplier:  Amarell
undefined
Thermometer, AMA freezer, red alcohol filled, measuring range:-35:20/0.5C, capillary yellow backed, dimension: 280x28mm, enclosed s cale, complete in plastic casing, rounded top finish, government verified with verification stamp. 1 * 1 items
Catalog # 620-1053
Supplier:  Amarell