ContactUsLinkComponent
Thermometer, precision, mercury filled, blue reflecting, measuring range:0:200/1°C, with mercury protection, PTFE coated, dimension : 305x8-9mm, enclosed scale, capillary lens front, total immersion, round richter-top finish, environmentally, suita 1 * 1 items
Catalog # 620-1213
Supplier:  Amarell
Thermometer, precision, mercury filled, blue reflecting, measuring range:0:200/1°C, with mercury protection, PTFE coated, dimension : 305x8-9mm, enclosed scale, capillary lens front, total immersion, round richter-top finish, environmentally, suita 1 * 1 items
Catalog # 620-1213
Supplier:  Amarell