ContactUsLinkComponent
Thermometer, maximum, sterilization, mercury filled, measuring range: 80:130/2C, enclosed scale, capillary lens front, colourless, dimension:60x7mm, pin-top finish, manufactured for reading after cooling down to 23C, suitable for government veri 1 * 1 items
  AMARL38605
 :  Amarell
Thermometer, maximum, sterilization, mercury filled, measuring range: 80:130/2C, enclosed scale, capillary lens front, colourless, dimension:60x7mm, pin-top finish, manufactured for reading after cooling down to 23C, suitable for government veri 1 * 1 items
  AMARL38605
 :  Amarell