ContactUsLinkComponent
Thermometer, maximum, for swinging, mercury filled, measuring range:-10:150/1C, enclosed scale, capillary lens front, colourless, d imension:260x7-8mm, Richter-top finish with button, manufactured for reading after cooling down to 23C, suitable f 1 * 1 items
Catalog # AMARL37036
Supplier:  Amarell
Thermometer, maximum, for swinging, mercury filled, measuring range:-10:150/1C, enclosed scale, capillary lens front, colourless, d imension:260x7-8mm, Richter-top finish with button, manufactured for reading after cooling down to 23C, suitable f 1 * 1 items
Catalog # AMARL37036
Supplier:  Amarell