Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Thermometer, pentane, -200:30/0.5c, 400mm, enclosed, scale, Din12781, colourless. 1 * 1 items
Catalog # AMARL33044
Supplier:
Amarell
Expand image
Thermometer, pentane, -200:30/0.5c, 400mm, enclosed, scale, Din12781, colourless. 1 * 1 items
Catalog # AMARL33044
Supplier:
Amarell