ContactUsLinkComponent
Thermometer, precision, mercury filled, DIN 12778, DIN 12775 and similar DIN standards , measuring range: -10:50/0.1C, colourless, milk glass scale, total immersion, length:420mm, enclosed scale, capillary lens front, government verified with veri 1 * 1 items
  620-1141
 :  Amarell
Thermometer, precision, mercury filled, DIN 12778, DIN 12775 and similar DIN standards , measuring range: -10:50/0.1C, colourless, milk glass scale, total immersion, length:420mm, enclosed scale, capillary lens front, government verified with veri 1 * 1 items
  620-1141
 :  Amarell