ContactUsLinkComponent
Thermometer, precision, mercury filled, DIN 12775 , measuring range: 0:150/0.5C, colourless, milk glass scale, total immersion, le n gth:350mm, enclosed scale, capillary lens front, government verified with verification certificate. Until 10th Octo 1 * 1 items
Catalog # 620-1132
Supplier:  Amarell
Thermometer, precision, mercury filled, DIN 12775 , measuring range: 0:150/0.5C, colourless, milk glass scale, total immersion, le n gth:350mm, enclosed scale, capillary lens front, government verified with verification certificate. Until 10th Octo 1 * 1 items
Catalog # 620-1132
Supplier:  Amarell