ContactUsLinkComponent
Thermometer, for soil depth, mercury filled, blue reflecting, DIN 58664, measuring range:-10:30/0.1C, milk glass scale, built-in l e ngth 500mm, complete with plastic guide tube, protecting cap and packing ring, suitable for government verification 1 * 1 items
Catalog # AMARK188450
Supplier:  Amarell
Thermometer, for soil depth, mercury filled, blue reflecting, DIN 58664, measuring range:-10:30/0.1C, milk glass scale, built-in l e ngth 500mm, complete with plastic guide tube, protecting cap and packing ring, suitable for government verification 1 * 1 items
Catalog # AMARK188450
Supplier:  Amarell