Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Alcoholometer, mercury filled, DIN12803, EUROPA classIII, measuring range: 90-95:0.1%vol, with thermometer, ranging:5:25/0.2C, leng th:330mm, suitable for government verification. 1 * 1 items
AMARH848256S
:
Amarell
Alcoholometer, mercury filled, DIN12803, EUROPA classIII, measuring range: 90-95:0.1%vol, with thermometer, ranging:5:25/0.2C, leng th:330mm, suitable for government verification. 1 * 1 items
AMARH848256S
:
Amarell