ContactUsLinkComponent
Alcoholometer, mercury filled, DIN12803, EUROPA classIII, measuring range: 30-35:0.1%vol, with thermometer, ranging:5:25/0.2C, leng th:330mm, suitable for government verification. 1 * 1 items
Catalog # AMARH848232S
Supplier:  Amarell
undefined
Alcoholometer, mercury filled, DIN12803, EUROPA classIII, measuring range: 30-35:0.1%vol, with thermometer, ranging:5:25/0.2C, leng th:330mm, suitable for government verification. 1 * 1 items
Catalog # AMARH848232S
Supplier:  Amarell