ContactUsLinkComponent
Areometer, density, measuring range:1.540-1.600:0.001g/cm , reference temperature:20°C, length:300mm, without thermometer. 1 * 1 items
Catalog # AMARH800162S
Supplier:  Amarell
undefined
Areometer, density, measuring range:1.540-1.600:0.001g/cm , reference temperature:20°C, length:300mm, without thermometer. 1 * 1 items
Catalog # AMARH800162S
Supplier:  Amarell