Thermometer, general purpose, blue special liquid, measuring range:- 10:250/1C, length:300, diameter:7-8mm, capillary lens front, enclosed scale. 1 * 1 items
Catalog # 620-1136
Thermometer, general purpose, blue special liquid, measuring range:- 10:250/1C, length:300, diameter:7-8mm, capillary lens front, enclosed scale. 1 * 1 items
Catalog # 620-1136