ContactUsLinkComponent
Thermometer, general purpose, mercury, measuring range:-10:360/1C, length:340, diameter:7-8mm, capillary lens front, enclosed scal e , colourless. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively inten 1 * 1 items
Catalog # 620-1084
Supplier:  Amarell
Thermometer, general purpose, mercury, measuring range:-10:360/1C, length:340, diameter:7-8mm, capillary lens front, enclosed scal e , colourless. Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively inten 1 * 1 items
Catalog # 620-1084
Supplier:  Amarell